Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167745.1 | NP_001161217.1 | 1 - 2 | 460 | 196 | |
NM_028459.2 | NP_082735.2 | 1 - 2 | 460 | 196 | ||
GenBank mRNA | AJ318416.1 | - | 1 - 2 | 178 | 196 | |
AK014104.1 | - | 1 - 2 | 454 | 196 | ||
AK140189.1 | - | 1 - 2 | 378 | 196 | ||
AK143969.1 | - | 1 - 2 | 441 | 196 | ||
AK144789.1 | - | 1 - 2 | 436 | 196 | ||
AK159052.1 | - | 1 - 2 | 412 | 196 | ||
BC019951.1 | - | 1 - 2 | 350 | 196 | ||
BC055045.1 | - | 1 - 2 | 436 | 6489610 | 196 | |
BC058642.1 | - | 1 - 2 | 460 | 196 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |