Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170866.1 | NP_001164337.1 | 1 - 2 | 103 | 91 | |
GenBank mRNA | AI893360.1 | - | 1 - 2 | 99 | 91 | |
BY258456.1 | - | 1 - 2 | 104 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |