Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009154.2 | NP_033180.2 | 11 - 12 | 1928 | 67 | |
GenBank mRNA | AK031231.1 | - | 11 - 12 | 1921 | 67 | |
AK043386.1 | - | 11 - 12 | 1640 | 67 | ||
AK046502.1 | - | 11 - 12 | 1704 | 67 | ||
AK141612.1 | - | 11 - 12 | 1872 | 67 | ||
AK143026.1 | - | 11 - 12 | 1939 | 67 | ||
AK157706.1 | - | 11 - 12 | 1911 | 67 | ||
BC065137.1 | - | 10 - 11 | 1397 | 30548165 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |