Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164805.1 | NP_001158277.1 | 12 - 13 | 3064 | 89 | |
GenBank mRNA | AK133324.1 | - | 7 - 8 | 1144 | 89 | |
AK173072.1 | - | 12 - 13 | 2835 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |