Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164805.1 | NP_001158277.1 | 1 - 2 | 460 | 60 | |
GenBank mRNA | AK173072.1 | - | 1 - 2 | 231 | 60 | |
BB656281.1 | - | 1 - 2 | 460 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |