Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164805.1 | NP_001158277.1 | 26 - 27 | 5066 | 68 | |
GenBank mRNA | AK133324.1 | - | 21 - 22 | 3146 | 68 | |
AK173072.1 | - | 26 - 27 | 4837 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |