Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163664.1 | NP_001157136.1 | 4 - 5 | 138 | 82 | |
NM_001163665.1 | NP_001157137.1 | 4 - 5 | 138 | 82 | ||
NM_001163669.1 | NP_001157141.1 | 4 - 5 | 138 | 82 | ||
GenBank mRNA | AA028290.1 | - | 4 - 5 | 118 | 82 | |
AI574395.1 | - | 4 - 5 | 132 | 82 | ||
BC003747.1 | - | 4 - 5 | 134 | 3594256 | 82 | |
L48990.1 | - | 3 - 4 | 56 | 82 | ||
L49467.1 | - | 4 - 5 | 138 | 82 | ||
L49470.1 | - | 4 - 5 | 138 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |