Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163669.1 | NP_001157141.1 | 6 - 7 | 165 | 60 | |
GenBank mRNA | AA028290.1 | - | 6 - 7 | 145 | 60 | |
L49470.1 | - | 6 - 7 | 165 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |