Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603214 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603212 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603213 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879782 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
592187 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
875173 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408269 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
298668 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
181588 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914409 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
802794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
725045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
285542 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588873 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613114 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
234437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729453 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
248585 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
353171 |
BC128268.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
353171 |
NM_146345.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
50045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
67881 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107839 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P126964 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P117803 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P41953 |
BC018248.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P41953 |
AK042389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P41953 |
AK041101.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P41953 |
AK020264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P41953 |
NM_030221.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603214 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603199 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603196 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603191 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603184 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603209 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603203 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603194 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603188 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603211 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603207 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603201 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603182 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603213 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603180 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603212 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603193 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603204 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603198 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25603186 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01659901 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06402734 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P126964 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06402689 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P117803 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P41953 |
BC018248.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P41953 |
AK042389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P41953 |
AK041101.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P41953 |
AK020264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P41953 |
NM_030221.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
BC018248.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK042389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK041101.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
AK020264.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P140350 |
NM_030221.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139395 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P126964 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P117803 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06472802 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06472803 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06472797 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06472801 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603212 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603214 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25603213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25602996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25602997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25602995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25602998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25602992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603196 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25603084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603196 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603173 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06367528 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06426273 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603207 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06472805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603195 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603193 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06472796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603057 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603194 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603212 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603169 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603058 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603211 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603061 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06426277 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603166 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603178 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603201 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05954073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06472798 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603184 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603198 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06472804 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603175 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603053 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06483612 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603203 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603182 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603188 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06621820 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06389258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06490930 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25603055 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06374361 |
- |
- |