Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100005899.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005899.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005899.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005908.mm.1 |
NM_001037764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005908.mm.1 |
NM_001037764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005908.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005908.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005908.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005909.mm.1 |
NM_001037764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005909.mm.1 |
NM_001037764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005909.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005909.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005909.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005902.mm.1 |
NM_001037764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005902.mm.1 |
NM_001037764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005902.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005912.mm.1 |
NM_001037764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005912.mm.1 |
NM_001037764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005912.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005912.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005912.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005900.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005900.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005900.mm.1 |
TC1100000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005904.mm.1 |
NM_001037764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005904.mm.1 |
NM_001037764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005904.mm.1 |
NM_009021.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005904.mm.1 |
NM_009021.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005904.mm.1 |
TC1100000745.mm.1 |
MTA TC |