Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164228.1 | NP_001157700.1 | 10 - 11 | 1517 | 63 | |
GenBank mRNA | AK050861.1 | - | 10 - 11 | 1434 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |