Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033318.3 | NP_001028490.2 | 10 - 11 | 1515 | 72 | |
GenBank mRNA | AK030709.1 | - | 10 - 11 | 1482 | 72 | |
AK145163.1 | - | 10 - 11 | 1513 | 72 | ||
AK154693.1 | - | 10 - 11 | 1517 | 72 | ||
AK163143.1 | - | 1535 | 72 | |||
AK172200.1 | - | 10 - 11 | 1517 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |