Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033318.3 | NP_001028490.2 | 8 - 9 | 1241 | 82 | |
GenBank mRNA | AK030709.1 | - | 8 - 9 | 1208 | 82 | |
AK145163.1 | - | 8 - 9 | 1239 | 82 | ||
AK154693.1 | - | 8 - 9 | 1243 | 82 | ||
AK163143.1 | - | 1261 | 82 | |||
AK172200.1 | - | 8 - 9 | 1243 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |