Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700019003.mm.1 |
TC0700002105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019003.mm.1 |
NM_001081370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019003.mm.1 |
NM_001081370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019003.mm.1 |
NM_001113373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019003.mm.1 |
NM_001113373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019001.mm.1 |
TC0700002105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019001.mm.1 |
NM_001081370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019001.mm.1 |
NM_001081370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018999.mm.1 |
TC0700002105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018999.mm.1 |
NM_001081370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018999.mm.1 |
NM_001081370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019000.mm.1 |
TC0700002105.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019000.mm.1 |
NM_001081370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019000.mm.1 |
NM_001081370.2 |
Refseq |