Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080444.4 | NP_536692.1 | 1 - 2 | 417 | 69 | |
GenBank mRNA | AF398971.1 | - | 1 - 2 | 446 | 69 | |
AK028538.1 | - | 1 - 2 | 417 | 69 | ||
AK052929.1 | - | 1 - 2 | 333 | 69 | ||
BC141311.1 | - | 1 - 2 | 408 | 9055923 | 69 | |
BC141313.1 | - | 1 - 2 | 408 | 9055925 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |