Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172606.2 | NP_766194.2 | 2 - 3 | 335 | 147 | |
GenBank mRNA | AK089827.1 | - | 2 - 3 | 337 | 147 | |
BU516606.1 | - | 2 - 3 | 351 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |