Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289546.1 | NP_001276475.1 | 12 - 13 | 1217 | 67 | |
NM_175007.2 | NP_778172.1 | 12 - 13 | 1217 | 67 | ||
GenBank mRNA | AK132168.1 | - | 12 - 13 | 1218 | 67 | |
BC024817.1 | - | 2 - 3 | 198 | 67 | ||
BC054718.1 | - | 12 - 13 | 1197 | 5686478 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |