Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btn2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289614.1 | NP_001276543.1 | 4 - 5 | 983 | 69 | |
NM_001289615.1 | NP_001276544.1 | 4 - 5 | 924 | 69 | ||
NM_175938.3 | NP_787952.2 | 4 - 5 | 983 | 69 | ||
GenBank mRNA | AK051061.1 | - | 4 - 5 | 970 | 69 | |
AY177686.1 | - | 4 - 5 | 966 | 69 | ||
BC139760.1 | - | 4 - 5 | 810 | 40058494 | 69 | |
BC141341.1 | - | 4 - 5 | 956 | 9055953 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |