Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081058.2 | NP_001074527.1 | 2 - 3 | 2324 | 96 | |
NM_027118.1 | NP_081394.1 | 2 - 3 | 2324 | 96 | ||
GenBank mRNA | AK035493.1 | - | 2 - 3 | 998 | 95 | |
AK083577.1 | - | 2 - 3 | 710 | 96 | ||
AK173265.1 | - | 2 - 3 | 2324 | 96 | ||
BC076619.1 | - | 3 - 4 | 779 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |