Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081058.2 | NP_001074527.1 | 6 - 7 | 2992 | 144 | |
NM_027118.1 | NP_081394.1 | 6 - 7 | 2992 | 144 | ||
GenBank mRNA | AK035493.1 | - | 6 - 7 | 1663 | 144 | |
AK173265.1 | - | 6 - 7 | 2992 | 144 | ||
BC076619.1 | - | 7 - 8 | 1447 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |