Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033264.3 | NP_001028436.2 | 15 - 16 | 1602 | 96 | |
NM_001285777.1 | NP_001272706.1 | 14 - 15 | 1522 | 96 | ||
NM_001285779.1 | NP_001272708.1 | 15 - 16 | 1631 | 96 | ||
GenBank mRNA | AK039618.1 | - | 14 - 15 | 1410 | 96 | |
AK165896.1 | - | 15 - 16 | 1548 | 96 | ||
AK168761.1 | - | 8 - 9 | 734 | 96 | ||
AK220231.1 | - | 15 - 16 | 1554 | 96 | ||
BC141115.1 | - | 15 - 16 | 1602 | 9055727 | 96 | |
EU770627.1 | - | 15 - 16 | 1540 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |