Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013930.4 | NP_038958.2 | 7 - 8 | 883 | 68 | |
GenBank mRNA | AJ224761.1 | - | 6 - 7 | 774 | 68 | |
AK044323.1 | - | 7 - 8 | 864 | 68 | ||
AK098141.1 | - | 7 - 8 | 824 | 68 | ||
AK136216.1 | - | 7 - 8 | 832 | 68 | ||
AK149400.1 | - | 7 - 8 | 808 | 68 | ||
BC005420.1 | - | 7 - 8 | 869 | 3600654 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |