Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080467.3 | NP_536715.3 | 9 - 10 | 1095 | 59 | |
GenBank mRNA | AB050903.1 | - | 8 - 9 | 821 | 59 | |
AF326316.1 | - | 9 - 10 | 889 | 59 | ||
AF435090.1 | - | 9 - 10 | 1083 | 59 | ||
AK128892.1 | - | 9 - 10 | 1095 | 59 | ||
BC046979.1 | - | 10 - 11 | 1117 | 6315485 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |