Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026496.4 | NP_080772.2 | 11 - 12 | 1840 | 84 | |
GenBank mRNA | AF411213.1 | - | 11 - 12 | 1573 | 84 | |
AK161180.1 | - | 4 - 5 | 521 | 84 | ||
BC004783.1 | - | 2 - 3 | 246 | 84 | ||
BC042575.1 | - | 6 - 7 | 673 | 84 | ||
BC055035.1 | - | 11 - 12 | 1629 | 6306106 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |