Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026496.4 | NP_080772.2 | 8 - 9 | 1440 | 95 | |
GenBank mRNA | AF411213.1 | - | 8 - 9 | 1173 | 95 | |
AK002633.1 | - | 8 - 9 | 1441 | 95 | ||
BC055035.1 | - | 8 - 9 | 1229 | 6306106 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |