Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008729.2 | NP_032755.2 | 10 - 11 | 2288 | 84 | |
GenBank mRNA | BC138260.1 | - | 10 - 11 | 1833 | 8861279 | 84 |
BC138261.1 | - | 10 - 11 | 1833 | 8861280 | 84 | |
BC145217.1 | - | 10 - 11 | 1830 | 9053768 | 84 | |
U90331.1 | - | 10 - 11 | 2291 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |