Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007572.2 | NP_001007573.1 | 2 - 3 | 280 | 64 | |
GenBank mRNA | AK085479.1 | - | 2 - 3 | 326 | 64 | |
BC110554.1 | - | 2 - 3 | 280 | 40047811 | 64 | |
BC110555.1 | - | 2 - 3 | 280 | 40047816 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |