Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177322.3 | NP_796296.1 | 2 - 3 | 317 | 63 | |
GenBank mRNA | BC036175.1 | - | 2 - 3 | 216 | 4989471 | 63 |
CJ066917.1 | - | 2 - 3 | 319 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |