Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028886.1 | NP_083162.1 | 12 - 13 | 1438 | 145 | |
GenBank mRNA | AK014976.1 | - | 12 - 13 | 1442 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |