Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183308.2 | NP_899131.1 | 6 - 7 | 789 | 130 | |
GenBank mRNA | AK007350.1 | - | 6 - 7 | 758 | 130 | |
AK146311.1 | - | 6 - 7 | 757 | 130 | ||
AK152351.1 | - | 6 - 7 | 740 | 130 | ||
AK171926.1 | - | 6 - 7 | 747 | 130 | ||
BC037140.1 | - | 6 - 7 | 731 | 5027719 | 130 | |
BC055896.1 | - | 6 - 7 | 789 | 3482584 | 130 | |
BC062200.1 | - | 6 - 7 | 780 | 6399484 | 130 | |
L48514.1 | - | 6 - 7 | 725 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |