Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177151.3 | NP_796125.2 | 5 - 6 | 842 | 60 | |
GenBank mRNA | AK049111.1 | - | 6 - 7 | 962 | 60 | |
BC158052.1 | - | 5 - 6 | 822 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |