Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177151.3 | NP_796125.2 | 58 - 59 | 11470 | 64 | |
GenBank mRNA | AK122302.1 | - | 19 - 20 | 4041 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |