Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289787.1 | NP_001276716.1 | 5 - 6 | 785 | 81 | |
NM_030727.5 | NP_109652.3 | 7 - 8 | 971 | 81 | ||
GenBank mRNA | AF529192.1 | - | 7 - 8 | 971 | 81 | |
AY024359.2 | - | 5 - 6 | 737 | 81 | ||
BC108987.1 | - | 5 - 6 | 777 | 40047725 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |