Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300010749.mm.1 |
NM_010093.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010749.mm.1 |
NM_010093.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010749.mm.1 |
TC1300001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010752.mm.1 |
NM_010093.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010752.mm.1 |
NM_010093.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010752.mm.1 |
TC1300001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010755.mm.1 |
NM_010093.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010755.mm.1 |
NM_010093.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010755.mm.1 |
TC1300001729.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010756.mm.1 |
NM_010093.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010756.mm.1 |
NM_010093.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010756.mm.1 |
TC1300001729.mm.1 |
MTA TC |