Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005554.1 | - | 1 - 2 | 277 | 82 | |
AK019929.1 | - | 1 - 2 | 358 | 82 | ||
AK030316.1 | - | 1 - 2 | 262 | 83 | ||
AK043342.1 | - | 1 - 2 | 335 | 82 | ||
AK162227.1 | - | 2 - 3 | 195 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |