Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014057.mm.1 |
TC0700001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014057.mm.1 |
NM_175645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014057.mm.1 |
NM_175645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014060.mm.1 |
TC0700001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014060.mm.1 |
NM_175645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014060.mm.1 |
NM_175645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014058.mm.1 |
TC0700001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014058.mm.1 |
NM_175645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014058.mm.1 |
NM_175645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014062.mm.1 |
TC0700001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014062.mm.1 |
NM_175645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014062.mm.1 |
NM_175645.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014059.mm.1 |
TC0700001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014059.mm.1 |
NM_175645.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014059.mm.1 |
NM_175645.3 |
Refseq |