Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101507.1 | NP_001094977.1 | 3 - 4 | 592 | 60 | |
GenBank mRNA | CJ059176.1 | - | 3 - 4 | 383 | 60 | |
CJ059317.1 | - | 2 - 3 | 190 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |