Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011539.3 | NP_035669.3 | 3 - 4 | 400 | 74 | |
GenBank mRNA | AK048544.1 | - | 3 - 4 | 382 | 74 | |
AK139479.1 | - | 3 - 4 | 363 | 74 | ||
BC029014.1 | - | 3 - 4 | 402 | 5007629 | 74 | |
L18868.1 | - | 3 - 4 | 271 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |