Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153600.2 | NP_705828.2 | 10 - 11 | 1018 | 97 | |
GenBank mRNA | AK035185.1 | - | 10 - 11 | 1026 | 97 | |
BC066060.1 | - | 10 - 11 | 1018 | 6848602 | 97 | |
BC066087.1 | - | 11 - 12 | 1208 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |