Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026662.4 | NP_080938.1 | 3 - 4 | 515 | 98 | |
GenBank mRNA | AK012819.1 | - | 3 - 4 | 547 | 98 | |
AK019169.2 | - | 3 - 4 | 484 | 98 | ||
AK029690.1 | - | 3 - 4 | 530 | 98 | ||
AK045007.1 | - | 3 - 4 | 516 | 98 | ||
BC024942.1 | - | 3 - 4 | 502 | 4022452 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |