Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010809.1 | NP_034939.1 | 1 - 2 | 184 | 74 | |
GenBank mRNA | AK148467.1 | - | 1 - 2 | 162 | 74 | |
BC006725.1 | - | 1 - 2 | 148 | 3962288 | 74 | |
X63162.1 | - | 1 - 2 | 184 | 74 | ||
X66402.1 | - | 1 - 2 | 126 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |