Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175484.3 | NP_780693.2 | 5 - 6 | 1021 | 82 | |
GenBank mRNA | AK047527.1 | - | 5 - 6 | 963 | 82 | |
AK087414.1 | - | 5 - 6 | 1022 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |