Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173038.3 | NP_766626.1 | 2 - 3 | 310 | 146 | |
GenBank mRNA | AK052261.1 | - | 2 - 3 | 279 | 146 | |
AK077017.1 | - | 2 - 3 | 312 | 146 | ||
AK084854.1 | - | 2 - 3 | 323 | 146 | ||
BC047462.1 | - | 2 - 3 | 177 | 146 | ||
BC139386.1 | - | 2 - 3 | 308 | 8862408 | 146 | |
BC139387.1 | - | 2 - 3 | 308 | 8862409 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |