Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900003512.mm.1 |
NM_001110826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003512.mm.1 |
NM_001110826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003512.mm.1 |
NM_181324.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003512.mm.1 |
NM_181324.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003512.mm.1 |
TC0900000515.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003513.mm.1 |
NM_001110826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003513.mm.1 |
NM_001110826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003513.mm.1 |
TC0900000515.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_001110826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_001110826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_007841.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_007841.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_181324.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
NM_181324.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003515.mm.1 |
TC0900000515.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_001110826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_001110826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_007841.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_007841.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_181324.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
NM_181324.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003511.mm.1 |
TC0900000515.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_001110826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_001110826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_007841.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_007841.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_181324.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
NM_181324.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003514.mm.1 |
TC0900000515.mm.1 |
MTA TC |