Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153537.4 | NP_705765.3 | 11 - 12 | 2759 | 85 | |
GenBank mRNA | AK122336.1 | - | 8 - 9 | 2477 | 85 | |
BC053931.1 | - | 7 - 8 | 1029 | 85 | ||
BC058712.1 | - | 11 - 12 | 2755 | 6842596 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |