Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016808.2 | NP_058088.2 | 7 - 8 | 1417 | 75 | |
NM_198091.2 | NP_932759.1 | 6 - 7 | 873 | 75 | ||
NM_198092.2 | NP_932760.2 | 7 - 8 | 1514 | 75 | ||
GenBank mRNA | AB041799.1 | - | 6 - 7 | 834 | 75 | |
AF079565.1 | - | 6 - 7 | 475 | 75 | ||
AK138799.1 | - | 8 - 9 | 1715 | 75 | ||
AY255637.1 | - | 6 - 7 | 842 | 75 | ||
AY255638.1 | - | 7 - 8 | 1396 | 75 | ||
AY255639.1 | - | 7 - 8 | 1350 | 75 | ||
AY255640.1 | - | 7 - 8 | 1395 | 75 | ||
BC017517.1 | - | 7 - 8 | 1396 | 4506362 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |