Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081412.2 | NP_001074881.1 | 18 - 19 | 3314 | 67 | |
GenBank mRNA | AK017213.1 | - | 3 - 4 | 373 | 67 | |
AK129482.1 | - | 15 - 16 | 1504 | 67 | ||
BC002193.1 | - | 1 - 2 | 109 | 67 | ||
BC048842.1 | - | 6 - 7 | 558 | 67 | ||
BC060270.1 | - | 18 - 19 | 2552 | 67 | ||
BC131682.1 | - | 19 - 20 | 2719 | 40111128 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |