Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007909.3 | NP_031935.3 | 1 - 2 | 406 | 71 | |
GenBank mRNA | AK140544.1 | - | 1 - 2 | 406 | 71 | |
BC048697.1 | - | 1 - 2 | 146 | 6510704 | 71 | |
U14752.1 | - | 1 - 2 | 179 | 71 | ||
U14941.1 | - | 1 - 2 | 134 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |