Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081055.1 | NP_001074524.1 | 6 - 7 | 931 | 74 | |
GenBank mRNA | AK154290.1 | - | 6 - 7 | 868 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |