Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030289.1 | NP_001025460.1 | 1 - 2 | 147 | 105 | |
GenBank mRNA | AK155813.1 | - | 1 - 2 | 212 | 105 | |
AK159182.1 | - | 1 - 2 | 143 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Mus musculus Probe spans exons |